Learn More
Trajan Scientific and Medical™ ProteCol™ PEEK Unions
Product Code. 15219213
Click to view available options
Description:
Size: 0.36 mm to 0.36 mm
Size: 1.59 mm to 0.36 mm
Size: 1.59 mm to 0.79 mm
Size: 1.59 mm to 1.59 mm
Unit Size:
Each
Pack of 2
Product Code. 15219213
Brand: Trajan Scientific and Medical™ 212011
Only null left
Add to basket
Add to basket
Precision engineered for capillary LC. Trajan Scientific and Medical ProteCol™ PEEK Unions offer complete versatility for connecting capillary tubing.
  • Leak free connections
  • Replacement parts such as nuts and ferrules
Specifications
Type Union
Description Size: 1.59 mm to 0.36 mm