missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 26S S5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
195.00€
Specifications
| Antigen | Proteasome 26S S5 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1.0 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18595684
|
Novus Biologicals
NB120-3319-0.025mg |
0.025 mg | |||||||
Description
Proteasome 26S S5 Polyclonal specifically detects Proteasome 26S S5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Proteasome 26S S5 | |
| Western Blot 1.0 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| PBS with 0.05% Sodium Azide | |
| 5711 | |
| Synthetic Peptide: L(485) S E G P Y Y V K P V S T T A V E G A E(504) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| KIAA007226S proteasome non-ATPase regulatory subunit 5, proteasome (prosome, macropain) 26S subunit, non-ATPase, 5,26S proteasome subunit S5B, S5BMGC23145,26S protease subunit S5 basic | |
| PSMD5 | |
| IgG | |
| Affinity Purified | |
| NB120-3319 immunizing peptide corresponds to amino acid residues 485-504 from human proteasome 19S subunit S5b protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title