missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 20S beta2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54589
This item is not returnable.
View return policy
Description
Proteasome 20S beta2 Polyclonal specifically detects Proteasome 20S beta2 in Human samples. It is validated for Western Blot.
Specifications
| Proteasome 20S beta2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 3.4.25.1, HC7-I, Macropain subunit C7-I, MGC104215, MGC126885, multicatalytic endopeptidase complex subunit C7-1, Multicatalytic endopeptidase complex subunit C7-I, proteasome (prosome, macropain) subunit, beta type, 2, proteasome beta 2 subunit, Proteasome component C7-I, proteasome subunit beta type-2, proteasome subunit, beta type, 2 | |
| Rabbit | |
| 23 kDa | |
| 100 μL | |
| Proteases & Other Enzymes | |
| 5690 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P49721 | |
| PSMB2 | |
| Synthetic peptides corresponding to PSMB2(proteasome (prosome, macropain) subunit, beta type, 2) The peptide sequence was selected from the middle region of PSMB2(NP_002785). Peptide sequence YDEHEGPALYYMDYLAALAKAPFAAHGYGAFLTLSILDRYYTPTISRERA. | |
| Affinity purified | |
| RUO | |
| Primary | |
| This product is specific to Subunit or Isoform: beta type-2. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction