missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 20S alpha 5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
195.00€
Specifications
| Antigen | Proteasome 20S alpha 5 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 μg/mL, Immunohistochemistry-Paraffin 1:10 - 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18528950
|
Novus Biologicals
NB120-11437-0.025mg |
0.025 mg | |||||||
Description
Proteasome 20S alpha 5 Polyclonal specifically detects Proteasome 20S alpha 5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Proteasome 20S alpha 5 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 μg/mL, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS containing 1mg/ml BSA with 0.05% Sodium Azide | |
| 5686 | |
| Synthetic peptide corresponding to residues C A(120) L Q F G E E D A D P G A M(133) of human proteasome 20S subunit alpha 5. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 3.4.25.1, FLJ42315, macropain subunit zeta, Macropain zeta chain, MGC117302, MGC125802, MGC125803, MGC125804, Multicatalytic endopeptidase complex zeta chain, proteasome (prosome, macropain) subunit, alpha type, 5, proteasome alpha 5 subunit, proteasome component 5, proteasome subunit alpha type-5, proteasome subunit zeta, Proteasome zeta chain, PSC5, ZETA | |
| PSMA5 | |
| IgG | |
| Affinity Purified | |
| This detects proteasome 20S subunit alpha 5 from human, rat and mouse cell lysates. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title