missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promazine Sulfone N-Oxide (Promazine N,S,S-Trioxide), Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Promazine Sulfone N-Oxide (Promazine N,S,S-Trioxide)
Specifications
Specifications
| Chemical Name or Material | Promazine Sulfone N-Oxide |
| CAS | 26928-74-5 |
| Analyte or Component Names | Promazine Sulfone N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H20 N2 O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | N,N-Dimethyl-10H-phenothiazine-10-propanamine N,5,5-Trioxide, Phenothiazine, 10-[3-(dimethylamino)propyl]-, N,5,5-trioxide (8CI) |
| InChI Formula | InChI=1S/C17H20N2O3S/c1-19(2,20)13-7-12-18-14-8-3-5-10-16(14)23(21,22)17-11-6-4-9-15(17)18/h3-6,8-11H,7,12-13H2,1-2H3 |
| SMILES | C[N+](C)([O-])CCCN1c2ccccc2S(=O)(=O)c3ccccc13 |
| IUPAC Name | 3-(5,5-dioxophenothiazin-10-yl)-N,N-dimethylpropan-1-amine oxide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction