missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prolactin Receptor Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148478
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor. Immunoreactive PRL receptor (PRLR), a member of the cytokine receptor family, varies in size (short and long forms) with tissue source and species, from ~40 kDa to 100 kDa. PRLR consists of at least three separate domains: an extracellular region with 5 cysteines which contains the prolactin binding site, a single transmembrane domain and a cytoplasmic region, the length of which appears to influence ligand binding and regulate cellular function.
Specifications
| Prolactin Receptor | |
| Polyclonal | |
| Unconjugated | |
| PRLR | |
| AI987712; cytokine receptor; HPRL; hPRL receptor; hPRLrI; Lactogen receptor; MFAB; pPRLR-SF; Pr-1; Pr-3; Prl R; PRLR; PRL-R; Prlr-rs1; Prolactin receptor; prolactin receptor 2 precursor; prolactin receptor related sequence 1; prolactin receptor short isoform; RATPRLR; secreted prolactin binding protein | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 5618 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P16471 | |
| PRLR | |
| Mouse myeloma cell line NS0-derived recombinant human Prolactin R, Gln25-Asp234. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction