missing translation for 'onlineSavingsMsg'
Learn More

Prolactin Rabbit anti-Human, Clone: PRL/4908R, Novus Biologicals™

Rabbit Monoclonal Antibody

517.00€

Specifications

Antigen Prolactin
Clone PRL/4908R
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1 to 2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18633517
View Documents
Novus Biologicals
NBP3-07241-100ug
100 μg
517.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Prolactin Monoclonal specifically detects Prolactin in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Prolactin
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction
10 mM PBS with 0.05% BSA
5617
IgG
Protein A or G purified
PRL/4908R
Immunohistochemistry-Paraffin 1 to 2 μg/mL
Monoclonal
Purified
RUO
Human
prolactin
Recombinant fragment of human Prolactin protein (around aa 63-201) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.