missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Prolactin R Antibody (BAY 1158061) - Humanized, Novus Biologicals™

Human Monoclonal Antibody

213.00€ - 920.00€

Specifications

Antigen Prolactin R
Clone BAY 1158061
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30047178
View Documents
Novus Biologicals™
NBP3-27967-50ug
50 μg
213.00€
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30044238
View Documents
Novus Biologicals™
NBP3-27967-1mg
1 mg
920.00€
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30044378
View Documents
Novus Biologicals™
NBP3-27967-100ug
100 μg
326.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Prolactin R Monoclonal antibody specifically detects Prolactin R in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

Prolactin R
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Apoptosis
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein
PRLR/ Prolactin Receptor
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
BAY 1158061
Monoclonal
Purified
RUO
Human
P16471
5618
IgG1
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.