missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proinsulin Antibody (CCI-17), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
564.00€
Specifications
| Antigen | Proinsulin |
|---|---|
| Clone | CCI-17 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Janelia Fluor 549 |
Description
Proinsulin Monoclonal antibody specifically detects Proinsulin in Human, Rat samples. It is validated for Western Blot, ELISA, ELISASpecifications
| Proinsulin | |
| Western Blot | |
| Janelia Fluor 549 | |
| Mouse | |
| Diabetes Research, Immune System Diseases, Immunology | |
| 50mM Sodium Borate | |
| 3630 | |
| IgG1 | |
| Protein A purified |
| CCI-17 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragment of rat proinsulin conjugated with a carrier protein. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title