missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prohibitin Antibody (S07-2G1), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00€ - 645.00€
Specifications
| Antigen | Prohibitin |
|---|---|
| Clone | S07-2G1 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645754
|
Novus Biologicals
NBP3-19909-100ul |
100 μg |
645.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663454
|
Novus Biologicals
NBP3-19909-50ul |
50 μg |
421.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prohibitin Monoclonal antibody specifically detects Prohibitin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Prohibitin | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 5245 | |
| IgG | |
| Affinity purified |
| S07-2G1 | |
| Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| PHB1, prohibitin | |
| A synthetic peptide of human Prohibitin (Uniprot # P35232) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title