missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Ensure accurate, reproducible results in ELISA, Radioimmunoassay
Progesterone Monoclonal specifically detects Progesterone in All Species samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Radioimmunoassay.
Specifications
Specifications
| Antigen | Progesterone |
| Applications | Radioimmune Assays (RIA), SDS-Page |
| Classification | Monoclonal |
| Clone | 6-5E-10B |
| Isotype | IgG1 κ |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?