missing translation for 'onlineSavingsMsg'
Learn More

Prodynorphin/PDYN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

326.00€ - 356.00€

Specifications

Antigen Prodynorphin/PDYN
Dilution Immunohistochemistry-Paraffin 1:50-1:200
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18664562
View Documents
Novus Biologicals
NBP2-97686-100ul
100 μg
326.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18614212
View Documents
Novus Biologicals
NBP2-97686-200ul
200 μg
356.00€
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Prodynorphin/PDYN Polyclonal antibody specifically detects Prodynorphin/PDYN in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Prodynorphin/PDYN
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Neuroscience
PBS (pH 7.0)
5173
IgG
Antigen and protein A Affinity-purified
Immunohistochemistry-Paraffin 1:50-1:200
Polyclonal
Purified
RUO
Human
ADCA, Beta-neoendorphin-dynorphin, leu-enkephalin, leumorphin, neoendorphin-dynorphin-enkephalin prepropeptide, PENKB, preprodynorphin, preproenkephalin B, proenkephalin-B, rimorphin, SCA23
Produced in rabbits immunized with E. coli-derived Human Prodynorphin/PDYN fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.