Learn More
Probenecid Methyl Ester, Mikromol™
Product Code. 30049533
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 30049533
Brand: Mikromol MM0657.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Probenecid Methyl Ester
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Probenecid Methyl Ester
CAS 53212-75-2
Analyte or Component Names Probenecid Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C14 H21 N O4 S
Certifications/Compliance ISO 17025
Synonym 4-[(Dipropylamino)sulfonyl]benzoic acid methyl ester
InChI Formula InChI=1S/C14H21NO4S/c1-4-10-15(11-5-2)20(17,18)13-8-6-12(7-9-13)14(16)19-3/h6-9H,4-5,10-11H2,1-3H3
SMILES CCCN(CCC)S(=O)(=O)c1ccc(cc1)C(=O)OC
IUPAC Name methyl 4-(dipropylsulfamoyl)benzoate
Molecular Weight (g/mol) 299.39
Quantity 25 mg
Formula Weight 299.1191 g/mol