missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pro-GDF1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148664
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Pro-GDF1, or pro-growth differentiation factor 1, is the precursor form of GDF1, a protein that belongs to the transforming growth factor-beta (TGF-beta) superfamily. GDF1 is involved in various developmental processes, particularly in embryogenesis and the development of the central nervous system and cardiovascular system. During its synthesis, pro-GDF1 undergoes proteolytic processing to become the mature, active form. GDF1 plays a crucial role in the formation of the left-right axis during embryonic development, which is essential for the proper positioning and orientation of internal organs. It is also implicated in the regulation of cell differentiation and growth. Mutations or dysregulation in the GDF1 gene can lead to congenital heart defects, heterotaxy (a condition where the internal organs are abnormally arranged), and other developmental anomalies. Understanding the function and regulation of pro-GDF1 and its mature form is important for elucidating the molecular mechanisms underlying normal development and congenital disorders. Research into pro-GDF1 provides insights into the broader roles of TGF-beta family proteins in growth and differentiation, with potential implications for therapeutic strategies aimed at correcting developmental defects and promoting tissue regeneration.
Specifications
| pro-GDF1 | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Mouse myeloma cell line NS0-derived recombinant mouse GDF-1 Propeptide, Pro25-Leu235. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction