missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pro-CRF Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149074
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Pro-CRF, or pro-corticotropin-releasing factor, is the precursor form of corticotropin-releasing factor (CRF), a crucial peptide hormone involved in the regulation of the hypothalamic-pituitary-adrenal (HPA) axis. The CRF gene encodes this precursor, which undergoes proteolytic processing to produce the mature CRF hormone. CRF plays a vital role in the body's response to stress by stimulating the release of adrenocorticotropic hormone (ACTH) from the anterior pituitary gland. ACTH subsequently promotes the secretion of cortisol from the adrenal cortex, which helps to manage stress and maintain homeostasis. Beyond its role in the stress response, CRF is also involved in various other physiological processes, including immune function, digestion, and energy balance. Dysregulation of CRF expression or signaling can lead to a range of disorders, such as depression, anxiety, and stress-related conditions, as well as metabolic and inflammatory diseases. Understanding the regulation, processing, and function of pro-CRF is essential for developing therapeutic strategies aimed at treating these conditions. Research into pro-CRF provides insights into the broader mechanisms of stress regulation and its impact on health and disease, highlighting its potential as a target for pharmacological intervention.
Specifications
| pro-CRF | |
| Polyclonal | |
| Unconjugated | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| E. coli-derived recombinant human Pro-Corticoliberin/Pro-CRH, Leu25-Lys196. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction