missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT6 Mouse anti-Human, Clone: PCRP-PRMT6-2C9, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07656-100ug
This item is not returnable.
View return policy
Description
PRMT6 Monoclonal specifically detects PRMT6 in Human samples. It is validated for Flow Cytometry, Protein Array.
Specifications
| PRMT6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1 to 2 μg/million cells, Protein Array | |
| EC 2.1.1.-, EC 2.1.1.125, FLJ10559, FLJ51477, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6, Histone-arginine N-methyltransferase PRMT6, HMT1 hnRNP methyltransferase-like 6, HMT1 hnRNP methyltransferase-like 6 (S. cerevisiae), HRMT1L6, protein arginine methyltransferase 6, protein arginine N-methyltransferase 6 | |
| Recombinant full-length human PRMT6 protein | |
| 100 μg | |
| Chromatin Research, Signal Transduction | |
| 55170 | |
| Store at 4°C. | |
| IgG2b |
| Flow Cytometry, Peptide Array | |
| PCRP-PRMT6-2C9 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction