missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKRIR Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95091-0.1ml
This item is not returnable.
View return policy
Description
PRKRIR Polyclonal antibody specifically detects PRKRIR in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PRKRIR | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 58 kDa interferon-induced protein kinase-interacting protein, DAP452 kDa repressor of the inhibitor of the protein kinase, Death-associated protein 4, inhibitor of protein kinase PKR, MGC102750, p52rIPK, p58IPK-interacting protein, protein-kinase, interferon-inducible double stranded RNA dependent inhibitor, repressor of (P58 repressor), THAP domain-containing protein 0, THAP0 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human THAP12 (NP_004696.2). MPNFCAAPNCTRKSTQSDLAFFRFPRDPARCQKWVENCRRADLEDKTPDQLNKHYRLCAKHFETSMICRTSPYRTVLRDNAIPTIFDLTSHLNNPHSRHRKRIKELSEDEIRTLKQKKIDETSEQEQKHKETNNSNAQNPSEEEGEGQDEDILPLTLEEKENKEYLKSLFEILILMGKQNIPLDGHEADEIPEGLFTPDNFQALLECRINSGEEVLRKRFETTAVNTLFCSKTQQRQMLEICESCIREETLREVRDSHFFSIITDDVVDIAGEEHLPVLVRFVDESHNLREEFIGFLPYE | |
| 0.1 mL | |
| Signal Transduction | |
| 5612 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction