missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PRKACB (Human) Recombinant Protein
Human PRKACB full-length ORF ( AAH16285, 1 a.a. - 257 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005567-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGNAATAKKGSEVESVKEFLAKAKEDFLKKWENPTQNNAGLEDFERKKTLGTGSFGRVMLVKHKATEQYYAMKILDKQKVVKLKQIEHTLNEKRILQAVNFPFLVRLEYAFKDNSNLYMVMEYVPGGEMFSHLRRIGRFSEPHARFYAAQIVLTFEYLHSLDLIYRDLKPENLLIDHQGYIQVTDFGFAKRVKGRTWTLCGTPEYLAPEIILSKGYNKAVDWWALGVLIYEMAAGYPPFFADQPIQIYEKIVSGKNF
Specifications
AAH16285 | |
protein kinase, cAMP-dependent, catalytic, beta | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PRKACB | |
GST |
5567 | |
Wheat germ expression system | |
10 μg | |
DKFZp781I2452, MGC41879, MGC9320, PKACB | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |