missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Presenilin-1 Antibody (APS 11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB300-749-50ug
This item is not returnable.
View return policy
Description
Presenilin-1 Monoclonal specifically detects Presenilin-1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Presenilin-1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:1000, ELISA 4.5 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 0.75 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 0.75 μg/mL | |
| AD3EC 3.4.23.-, Alzheimer disease 3, EC 3.4.23, FAD, presenilin 1, Protein S182, PS-1, PS1presenilin-1, PSNL1, S182 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 5663 | |
| Human | |
| Purified |
| Western Blot | |
| APS 11 | |
| Unconjugated | |
| P49768 | |
| PSEN1 | |
| Synthetic peptide corresponding to residues C H(21) L S N T V R S Q N D N R E(34) of human PS1. | |
| 50 μg | |
| Primary | |
| No cross-reactivity is seen with presenilin 2. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction