Learn More
Thermo Scientific™ Preparative HPLC Guard Column Holders
Product Code. 16358636 Shop All Thermo Scientific Products
Click to view available options
Column Format:
Guard Holder 1/16 in.
Guard Holder 1/8 in.
Guard Holder, Stand-Alone
Diameter (Metric):
10 mm
20 mm
Length (Metric):
8 mm
20 mm
30 mm
Unit Size:
Each
Product Code. 16358636
Brand: Thermo Scientific™ 889020
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Protect your Thermo Scientific Preparative HPLC Column from unintended particulates that may reduce the lifetime of your column by using a guard column and guard holder.
Using preparative guards is a proven and cost effective way to extend the lifetime of your preparative columns. Thermo Scientific™ Preparative Guard Column Holders allow for easy assembly and connection of the guard column to the preparative column.
Selecting the appropriate preparative guard holder for your size of preparative HPLC column is important. We offer a range of guard holder sizes which are all in a convenient stand-alone format that requires connections from the LC inlet and outlet to the preparative LC column.
Our preparative guard holders are available in 1/16 in. or 1/8 in. connection fittings in the larger guard sizes to be compatible with your Preparative HPLC system hardware.
TRUSTED_SUSTAINABILITY
Specifications
Column Format Guard Holder, Stand-Alone
Diameter (Metric) 20 mm
For Use With Preparative guard column
Length (Metric) 20 mm
Product Line Thermo Scientific
Type Guard Column Holder