Learn More
Eppendorf PremiumCare Services
Product Code. 32215722 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
CellXpert 170(i)
CellXpert CS220
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
32215722 CellXpert CS220 Each
30130785 CellXpert 170(i) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32215722 Supplier Eppendorf Supplier No. 0082050321
 Restricted Item
Estimated Shipment: 16-11-2026
A full annual preventive maintenance service program, unlimited travel, spare parts and labor on repairs included in the agreement; 10% discount on additional services (e.g. IQ/OQ) during the agreement period.
  • Include annual maintenance with:
    • Check of installation environment and electrical/gas supply
    • Inspection of equipment-specific sensors, doors/lids and key components
    • Validation and adjustment of operating parameters
  • Equipment-specific differences:
    • Multipurpose Centrifuges:
      • Housing and rotor cleaning
      • Checks of condensation tray, lid latch, display, keypad, stability, rotor/imbalance detection, temperature and speed sensors
    • New Brunswick S41i:
      • CO2 and auto-zero filter replacement
      • Checks of shaker platform, drive belt, humidity tray and support seals
    • CellXpert CS220:
      • CO2 filter replacement; checks of shaker platform, drive belt and rH sensor
    • CellXpert C 170 (i):
      • Gas and auto-zero filter replacement; checks of door, alarm, humidity tray, wiring, PCB, display and controller
    • Biological Shakers:
      • Door/lid hardware checks
      • Lubrication of shaker mechanism
      • Checks of drive motor and belt
      • Condenser cleaning and inspection
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CellXpert CS220
Product Type Service Plan