missing translation for 'onlineSavingsMsg'
Learn More

PRELID2 Monoclonal Antibody (OTI2F6), TrueMAB™, OriGene

Product Code. 17544204
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17544204 100 μL 100µL
17564204 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17544204 Supplier Origene Technologies Supplier No. TA810771

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRELID2 (PRELI Domain Containing 2) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include phosphatidic acid transporter activity. An important paralog of this gene is PRELID3B.

Specifications

Antigen PRELID2
Applications Western Blot
Classification Monoclonal
Clone OTI2F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PRELID2
Gene Accession No. Q8N945
Gene Alias PRELI domain containing 2
Gene Symbols PRELID2
Host Species Mouse
Immunogen Full length human recombit protein of human PRELID2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153768
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.