Learn More
Prednisolone Hemisuccinate, Mikromol™
Description
Prednisolone Hemisuccinate
Specifications
Specifications
| Chemical Name or Material | Prednisolone hemisuccinate |
| CAS | 2920-86-7 |
| Analyte or Component Names | Prednisolone hemisuccinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H32 O8 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (11beta)-21-(3-Carboxy-1-oxopropoxy)-11,17-dihydroxypregna-1,4-diene-3,20-dione, 11beta,17,21-Trihydroxypregna-1,4-diene-3,20-dione 21-(hydrogen succinate), Prednisolone 21-O-beta-carbonylpropionic acid, Prednisolone 21-hemisuccinate, Prednisolone 21-succinate, Prednisolone bisuccinate, Prednisolone hemisuccinate, Prednisolone succinate, Prednisolut, delta1-Hydrocortisone 21-hemisuccinate, Methylprednisolone Hydrogen Succinate Impurity E |
| InChI Formula | InChI=1S/C25H32O8/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30/h7,9,11,16-18,22,27,32H,3-6,8,10,12-13H2,1-2H3,(H,29,30)/t16-,17-,18-,22+,23-,24-,25-/m0/s1 |
| SMILES | C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)COC(=O)CCC(=O)O |
| IUPAC Name | 4-[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoic acid |
| Show More |