missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prednienic Acid, Mikromol™
Description
Prednienic Acid
Specifications
Specifications
| Chemical Name or Material | Prednienic Acid |
| CAS | 37927-29-0 |
| Analyte or Component Names | Prednienic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H26 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Prednienic Acid |
| InChI Formula | InChI=1S/C20H26O5/c1-18-7-5-12(21)9-11(18)3-4-13-14-6-8-20(25,17(23)24)19(14,2)10-15(22)16(13)18/h5,7,9,13-16,22,25H,3-4,6,8,10H2,1-2H3,(H,23,24)/t13-,14-,15-,16+,18-,19-,20-/m0/s1 |
| SMILES | C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)O |
| IUPAC Name | (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-17-carboxylic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?