missing translation for 'onlineSavingsMsg'
Learn More

PREB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

195.00€

Specifications

Antigen PREB
Dilution Western Blot 1:500
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18584673
View Documents
Novus Biologicals
NB120-3422-0.025ml
0.025 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

PREB Polyclonal specifically detects PREB in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

PREB
Western Blot
Unconjugated
Rabbit
Transcription Factors and Regulators
PBS with 0.05% Sodium Azide
10113
Recombinant mouse PREB.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Western Blot 1:500
Polyclonal
Purified
RUO
Mouse
MGC3467, prolactin regulatory binding-element protein, prolactin regulatory element binding, prolactin regulatory element-binding protein, SEC12Mammalian guanine nucleotide exchange factor mSec12
PREB
IgG
Ammonium sulfate precipitation
Detects recombinant mouse PREB (Prolactin Regulatory Element Binding protein).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.