Learn More
Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil
Product Code. 10013984 Shop All Thermo Scientific Products
Click to view available options
Diameter (English) Inner:
0.003 in.
0.005 in.
0.007 in.
0.010 in.
0.020 in.
0.030 in.
0.040 in.
Unit Size:
Each
Product Code. 10013984
Brand: Thermo Scientific™ 37010
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil is easily cut to the desired length.
Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil is easily cut to the desired length.
  • Broad chemical compatibility
  • Biocompatible
  • May be used with standard fingertight fittings
  • Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
TRUSTED_SUSTAINABILITY
Specifications
For Use With HPLC
Type HPLC Tubing
Description 0.010 in. ID; 5 ft. length; Blμe
For Use With (Application) HPLC
Diameter (English) Inner 0.010 in.
Diameter (Metric) Inner 0.25 mm
Overall Length (English) 5 ft.
Overall Length (Metric) 1.524 m
Material PEEK