missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDX3 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 81833-1-RR-20UL
This item is not returnable.
View return policy
Description
The PRDX3 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human PRDX3. This antibody recognizes human, mouse, rat antigen. The PRDX3 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| PRDX3 | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| AOP-1; AOP1; MER5; MGC104387; MGC24293; PRO1748; SP-22 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 28 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1L11 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 10935 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu