missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM4 Antibody (RAB-C367) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
244.00€ - 635.00€
Specifications
| Antigen | PRDM4 |
|---|---|
| Clone | RAB-C367 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18689004
|
Novus Biologicals
NBP3-20139-0.025mg |
0.025 mg |
244.00€
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644784
|
Novus Biologicals
NBP3-20139-0.2mg |
0.2 mg |
635.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRDM4 Monoclonal antibody specifically detects PRDM4 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)Specifications
| PRDM4 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC45046, PFM1PR domain-containing protein 4, PR domain containing 4, PR domain zinc finger protein 4, PR-domain zinc-finger protein PFM1 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM4 protein under non-denaturing conditions. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| RAB-C367 | |
| Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research | |
| PBS | |
| 11108 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title