missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRCP Monoclonal Antibody (860821)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561772
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Cleaves C-terminal amino acids linked to proline in peptides such as angiotensin II, III and des-Arg9-bradykinin. This cleavage occurs at acidic pH, but enzymatic activity is retained with some substrates at neutral pH.
Specifications
| PRCP | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P42785 | |
| PRCP | |
| Mouse myeloma cell line NS0-derived recombinant human Lysosomal Pro-X Carboxypeptidase/PRCP, Met1-His496. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 860821 | |
| Unconjugated | |
| PRCP | |
| 2510048K03Rik; 2610104A14Rik; AI451719; Angiotensinase C; HUMPCP; Lysosomal carboxypeptidase C; lysosomal Pro-X carboxypeptidase; PCP; Prcp; Proline carboxypeptidase; prolylcarboxypeptidase; prolylcarboxypeptidase (angiotensinase C) | |
| Rat | |
| Protein A/G | |
| RUO | |
| 5547 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction