missing translation for 'onlineSavingsMsg'
Learn More

Pravastatin Methyl Ester, Mikromol™

Product Code. 30049870
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30049870

Brand: Mikromol MM0503.090025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pravastatin Methyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pravastatin Methyl Ester
CAS 81131-72-8
Analyte or Component Names Pravastatin Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C24 H38 O7
Certifications/Compliance ISO 9001
Synonym (betaR,deltaR,1S,2S,6S,8S,8aR)-1,2,6,7,8,8a-hexahydro-beta,delta,6-trihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-1-Naphthaleneheptanoic acid methyl ester, 1-Naphthaleneheptanoic acid, 1,2,6,7,8,8a-hexahydro-beta,delta,6-trihydroxy-2-methyl-8-(2-methyl-1-oxobutoxy)-, methyl ester, [1S-[1alpha(betaS*,deltaS*),2alpha,6alpha,8beta(R*),8aalpha]]- (9CI)
InChI Formula InChI=1S/C24H38O7/c1-5-14(2)24(29)31-21-12-18(26)10-16-7-6-15(3)20(23(16)21)9-8-17(25)11-19(27)13-22(28)30-4/h6-7,10,14-15,17-21,23,25-27H,5,8-9,11-13H2,1-4H3/t14-,15-,17+,18+,19+,20-,21-,23-/m0/s1
SMILES CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)OC)[C@@H]12
IUPAC Name methyl (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-[(2S)-2-methylbutanoyl]oxy-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
Molecular Weight (g/mol) 438.55
Quantity 25 mg
Formula Weight 438.2618 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.