missing translation for 'onlineSavingsMsg'
Learn More

PRAMEF20 Antibody - BSA Free, Novus Biologicals™

Product Code. 18610519 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18610519 100 μg 100µL
18671858 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18610519 Supplier Novus Biologicals Supplier No. NBP305596100ul

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRAMEF20 Polyclonal antibody specifically detects PRAMEF20 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRAMEF20
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias PRAME family member 20, PRAME family member 20/21, PRAME family member 21, PRAMEF21
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-477 of mouse PRAMEF20 (NP_001108549.1). MSSKPLLTLQELAAQNILRNEALAISALQYLPELFLPQLLKQAYDGNHLKVLRAIVSSWPFPRLPLGALKKRTPYLETQLQVVLEEVDKLLIQEVHPREYKLEVLDLRSVGQNYLNVWPGAMDDWLPKTTQTDPCCSKTVMTQPLKVLVDLQLNNRGQSRLFSYLVQWSDRRKGLLQLYCNKLQIWSPSQQNYRKLSQKVNLDYVETLGLHSSCSPSFLLNFAPYLRQMRNLRKLALSNIREEHIISPEKRRCIITRFTLQILKLESLQKLHLDAVCFLEGHLDQLLGSVKTSLDELAVTHCNLSVSEWSHFSEFPCVSQLQQLNLENVRLTGLSPEPLQVLLVKAGPTLVALDLEDCHLEDGQLYAILPALSKCFKLTKFSFYGNQISMLAMKDLLHHTASLIHLRLELYPVPQDSYDYHGTPLMQRMQQHCDELMNLLKTFRNPGRVFFGTDRCDQCCNRYIYNKTIMCNCRRSY
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 645425
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.