Learn More
PRAME Antibody (CL5146) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61148H
Product Code. 30289304
  • 737.00€ / 0.10mL
Description
PRAME Monoclonal antibody specifically detects PRAME in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRAME
Monoclonal
HRP
cancer/testis antigen 130, MAPEOIP4CT130, melanoma antigen preferentially expressed in tumors, OIP-4, Opa-interacting protein 4, Opa-interacting protein OIP4, preferentially expressed antigen in melanoma, Preferentially expressed antigen of melanoma
This antibody was developed using a recombinant protein derived from P78395, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
23532
Store at 4⪚C in the dark.
IgG2a
Western Blot, Immunocytochemistry/Immunofluorescence
CL5146
PBS
Mouse
Protein A purified