missing translation for 'onlineSavingsMsg'
Learn More

PR3 (Proteinase 3) Ab-1, Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ PR3 (Proteinase 3) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-1333-PABX

 View more versions of this product

Product Code. 12619596

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

This item is not returnable. View return policy

Description

Description

Proteinase 3 (PR3), a constituent of azurophil granules of neutrophils (polymorphonuclear cells, PMNs), is the target antigen for most anti-neutrophil cytoplasmic antibodies (c-ANCA) in Wegener's granulomatosis (WG).

Host Species: Mouse

Clone: MCPR3-2

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: HMC-1/PR3 cell granule extract

Molecular Weight: 32-40kDa

Positive Control: HMC-1/PR3-S176 cells

Recommended for:

  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PR3 (Proteinase 3) Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Proteases And Inhibitors
Human
IgG2a κ
Immunofluorescence
MCPR3-2
Unconjugated
HMC-1/PR3 cell granule extract
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.