missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPP4R1 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
213.00€ - 508.00€
Specifications
| Antigen | PPP4R1 |
|---|---|
| Dilution | Western Blot : 5-10 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066842
|
Novus Biologicals™
NBP3-30784-100ug |
100 μg |
508.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067093
|
Novus Biologicals™
NBP3-30784-25ug |
25 μg |
213.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPP4R1 Polyclonal antibody specifically detects PPP4R1 in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
| PPP4R1 | |
| Western Blot | |
| Unconjugated | |
| Sheep | |
| Protein Phosphatase | |
| Phosphate buffered saline | |
| 9989 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 5-10 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| MEG1, PP4R1PP4(Rmeg), protein phosphatase 4, regulatory subunit 1, serine/threonine phosphatase 4, serine/threonine-protein phosphatase 4 regulatory subunit 1 | |
| Antibody raised against synthetic peptide corresponding to amino acids 38 to 52 of the human protein PPP4R1. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title