Learn More
Novus Biologicals™ PPP4R1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30785-100ug
This item is not returnable.
View return policy
Description
PPP4R1 Polyclonal antibody specifically detects PPP4R1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| PPP4R1 | |
| Polyclonal | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| MEG1, PP4R1PP4(Rmeg), protein phosphatase 4, regulatory subunit 1, serine/threonine phosphatase 4, serine/threonine-protein phosphatase 4 regulatory subunit 1 | |
| Antibody raised against synthetic peptide corresponding to amino acids 38 to 52 of the human protein PPP4R1. | |
| 100 μg | |
| Protein Phosphatase | |
| 9989 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |