missing translation for 'onlineSavingsMsg'
Learn More

PPP3R2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen PPP3R2
Concentration 0.25 mg/mL
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16872464
View Documents
Proteintech
14005-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16862464
View Documents
Proteintech
14005-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PPP3R2 is regulatory subunit of calcineurin, a calcium-dependent, calmodulin stimulated protein phosphatase. This protein confers calcium sensitivity.
Specifications

Specifications

PPP3R2
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse
Q63811, Q96LZ3
19059, 5535
PPP3R2 Fusion Protein Ag5097
Primary
-20°C
PPP3R2
0.25 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Calcineurin B like protein, Calcineurin BII, Calcineurin subunit B type 2, CBLP, CNBII, PPP3R2, PPP3RL
Ppp3r2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.