Learn More
PPP3CC Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15582-1-AP-20UL
Additional Details : Weight : 0.00020kg
Description
The PPP3CC antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human PPP3CC. This antibody recognizes human, mouse, rat antigen. The PPP3CC antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
PPP3CC | |
Polyclonal | |
Unconjugated | |
20 μL | |
5533 | |
Liquid |
Western Blot, ELISA | |
240 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse, Rat |