Learn More
Abnova™ PPP2R2B (Human) Recombinant Protein
Product Code. 16084545
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16084545
Brand: Abnova™ H00005521Q02.10ug
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Human PPP2R2B partial ORF ( AAH31790.1, 22 a.a. - 128 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: TEADIISTVEFNHTGELLATGDKGGRVVIFQREQESKNQVHRRGEYNVYSTFQSHEPEFDYLKSLEIEEKINKIRWLPQQNAAYFLLSTNDKTVKLWKVSERDKRPE
Specifications
Accession Number AAH31790.1
Gene ID (Entrez) 5521
Name protein phosphatase 2 (formerly 2A), regulatory subunit B, beta isoform
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias B55-BETA, FLJ95686, MGC24888, PP2A-B55BETA, PP2A-PR55B, PP2AB-BETA, PP2APR55-BETA, PR2AB-BETA, PR2AB55-BETA, PR2APR55-BETA, PR52B, PR55-BETA, SCA12
Gene Symbol PPP2R2B
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer