missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R9B/Spinophilin Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94055-0.02ml
This item is not returnable.
View return policy
Description
PPP1R9B/Spinophilin Polyclonal antibody specifically detects PPP1R9B/Spinophilin in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PPP1R9B/Spinophilin | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| FLJ30345, neurabin II, neurabin-2, neurabin-II, PPP1R6, PPP1R9, Protein phosphatase 1 regulatory subunit 9B, protein phosphatase 1, regulatory (inhibitor) subunit 9B, protein phosphatase 1, regulatory subunit 9B, spinophilin, SPINO, spinophilin, Spn | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human PPP1R9B/Spinophilin (NP_115984.3). VFQPPPPPPPAPSGDAPAEKERCPAGQQPPQHRVAPARPPPKPREVRKIKPVEVEESGESEAESAPGEVIQAEVTVHAALENGSTVATAASPAPEEPKAQA | |
| 0.02 mL | |
| Cell Cycle and Replication, Neuroscience | |
| 84687 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction