missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP1R2 Monoclonal Antibody (488123)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561478

Product Code. 30534334

  • 429.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Protein phosphatase-1 (PP1) is one of the main eukaryotic serine/threonine phosphatases. The protein encoded by this gene binds to the catalytic subunit of PP1, strongly inhibiting its activity. Ten related pseudogenes have been found throughout the human genome. Several splice variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PPP1R2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P41236
PPP1R2
E. coli-derived recombinant human PP1 Inhibitor-2, Met1-Ser205.
100 μg
Primary
Human
Antibody
IgG2b
Western Blot
488123
Unconjugated
PPP1R2
0610025N14Rik; 2310007G06Rik; 4930440J04Rik; 5430408E15Rik; D16Ertd248e; Ipp2; IPP-2; phosphatase inhibitor 2; phosphoprotein phosphatase; PPP1R2; protein phosphatase 1 regulatory inhibitor subunit 2; protein phosphatase 1, regulatory (inhibitor) subunit 2; protein phosphatase 1, regulatory subunit 2; protein phosphatase inhibitor 2
Mouse
Protein A/G
RUO
5504
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.