missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAR delta/NR1C2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
200.00€
Specifications
| Antigen | PPAR delta/NR1C2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:250 - 1:750, Flow Cytometry 3 - 5 ug 106 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 2 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
Description
PPAR delta/NR1C2 Polyclonal specifically detects PPAR delta/NR1C2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| PPAR delta/NR1C2 | |
| Western Blot 1:250 - 1:750, Flow Cytometry 3 - 5 ug 106 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 2 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| FAAR, MGC3931, NR1C2PPAR-delta, NUC1PPAR-beta, NUCI, NUCII, Nuclear hormone receptor 1, Nuclear receptor subfamily 1 group C member 2, peroxisome proliferative activated receptor, delta, Peroxisome proliferator-activated receptor beta, peroxisome proliferator-activated receptor delta, PPARB | |
| PPARD | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| 1 mg/mL BSA with 0.05% Sodium Azide | |
| 5467 | |
| Synthetic peptide corresponding to residues M(1) E Q P Q E E T P E A R E E(14) C of mouse PPAR delta. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title