missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PP5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

213.00€ - 508.00€

Specifications

Antigen PP5
Dilution Western Blot : 5-10 μg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30067020
View Documents
Novus Biologicals™
NBP3-30838-25ug
25 μg
213.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30067244
View Documents
Novus Biologicals™
NBP3-30838-100ug
100 μg
508.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PP5 Polyclonal antibody specifically detects PP5 in Human samples. It is validated for Western Blot
Specifications

Specifications

PP5
Western Blot
Unconjugated
Rabbit
Cell Cycle and Replication
Phosphate buffered saline
5536
IgG
Ammonium sulfate precipitation
Western Blot : 5-10 μg/mL
Polyclonal
Purified
RUO
Human
EC 3.1.3.16, PP5FLJ36922, PPP5, PPT, PP-T, protein phosphatase 5, catalytic subunit, Protein phosphatase T, Serine/Threonine phosphatase, serine/threonine-protein phosphatase 5
Rabbits were immunized with a Synthetic peptide derived from the human PP5
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.