missing translation for 'onlineSavingsMsg'
Learn More

PP4/PPP4C Antibody, Novus Biologicals™

Product Code. 18292750 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18292750 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18292750 Supplier Novus Biologicals Supplier No. NBP131117

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PP4/PPP4C Polyclonal specifically detects PP4/PPP4C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP4C
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.45mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias EC 3.1.3.16, Pp4, PP4C, PP4protein phosphatase X, catalytic subunit, PPP4, PP-X, PPXPPH3, protein phosphatase 4 (formerly X), catalytic subunit, protein phosphatase 4, catalytic subunit, Protein phosphatase X, serine/threonine-protein phosphatase 4 catalytic subunit
Gene Symbols PPP4C
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human PPP4C. The exact sequence is proprietary.
Molecular Weight of Antigen 35 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Protein Phosphatase, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5531
Test Specificity Tested in human. Predicted cross-reactivity based on sequence identity: Xenopus laevis (93%), Xenopus tropicalis (93%), and Rhesus Monkey (100%). Not yet tested in any other species.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.