Learn More
Novus Biologicals™ VAV2 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30790-250ug
This item is not returnable.
View return policy
Description
VAV2 Polyclonal antibody specifically detects VAV2 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| VAV2 | |
| Polyclonal | |
| Western Blot : 1:5000, Immunoprecipitation | |
| guanine nucleotide exchange factor VAV2, oncogene VAV2, Protein vav-2, vav 2 guanine nucleotide exchange factor, vav 2 oncogene, VAV-2 | |
| A his tagged fusion protein VAV2 corresponding to amino acid sequence 578 - 878 and having a molecular weight of 100kD | |
| 250 μg | |
| Signal Transduction | |
| 7410 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |