missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PP1 alpha/PPP1A Antibody, Novus Biologicals™

Product Code. 30066867 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30066867 100 μg 100µg
30067000 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30066867 Supplier Novus Biologicals™ Supplier No. NBP330764100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PP1 alpha/PPP1A Polyclonal antibody specifically detects PP1 alpha/PPP1A in Bovine,Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP1 alpha/PPP1A
Applications Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot : 1:500-1:1000, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation Phosphate buffered saline
Gene Alias EC 3.1.3.16, PP1alpha, PP-1AMGC15877, PPP1AMGC1674, protein phosphatase 1, catalytic subunit, alpha isoform, protein phosphatase 1, catalytic subunit, alpha isozyme, serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit, serine/threonine-protein phosphatase PP1-alpha catalytic subunit
Host Species Rabbit
Immunogen The immunogen used to generate the purified antibody was a peptide conjugated to KLH corresponding to the sequence NH2-Arg-Pro-Ile-Thr-Pro-Pro-Arg-Asn-Ser-AlaLys-Ala-Lys-Lys-COOH. This peptide antibody corresponds to C -terminal peptide of PP1 alpha/PPP1A catalytic subunit having a MW of 38kDa. The sequence used is amino acid 317-330.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5499
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.