missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU4F2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | POU4F2 |
|---|---|
| Concentration | 280 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234803
|
Proteintech
55042-1-AP-150UL |
150 μL |
410.00€ 349.00€ / 150µL Save 61.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234570
|
Proteintech
55042-1-AP-20UL |
20 μL |
145.00€ 129.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The POU4F2 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human POU4F2. This antibody recognizes human, mouse antigen. The POU4F2 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| POU4F2 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| BRN3.2; BRN3B; Brn-3b | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 280 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 5458 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title