Learn More
Potassium Standard Solution, 1000μg/mL, Spectro ECON, Chem-Lab
Product Code. 11978504
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11978504 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11978504 Supplier Chem-Lab Supplier No. CL01.1105.0500
  • ONLINE EXCLUSIVE PRICE
    43.00€
    25.50€ / 500mL
    Save 17.50€
    40% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Used as a reference standard in spectroscopy and other laboratory applications. Potassium Standard Solution, 1000μg/mL, Spectro ECON, Chem-Lab is a mixture of 1.9g KCl per liter, to form a solution containing 1mg/mL of K1+.
This potassium standard solution contains a precise concentration of 990–1010μg/mL of K1+ ±0.5%. It is used to determine the concentration of solutions in titrations. Potassium standard has been used in atomic absorption spectroscopy.
Specifications
CAS 7447-40-7, 7732-18-5
Density 1.00g/mL
Physical Form Liquid
Molecular Formula ClK
Concentration 990–1010 μg/mL K1+ (±0.5%)
MDL Number MFCD00011360
Color Colorless
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium chloride
Molecular Weight (g/mol) 74.55
PubChem CID 4873
Quantity 500 mL
ChEBI CHEBI:32588
Formula Weight 74.56g/mol
Concentration or Composition (by Analyte or Components) 1.907g KCl per 1L H2O