missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Potassium Channel Kv3.1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30824-25ug
This item is not returnable.
View return policy
Description
Potassium Channel Kv3.1 Polyclonal antibody specifically detects Potassium Channel Kv3.1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| Potassium Channel Kv3.1 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| FLJ41162, FLJ42249, FLJ43491, KV3.1, KV4, MGC129855, NGK2, potassium voltage-gated channel subfamily C member 1, potassium voltage-gated channel, Shaw-related subfamily, member 1, voltage-gated potassium channel protein KV3.1, Voltage-gated potassium channel subunit Kv3.1, Voltage-gated potassium channel subunit Kv4 | |
| Rabbits were immunized with a synthetic peptide derived from the rat Potassium Channel Kv3.1 conjugated to KLH | |
| 25 μg | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Potassium Channels | |
| 3746 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction