missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Porimin Monoclonal antibody specifically detects Porimin in Human samples. It is validated for Immunohistochemistry
Specifications
Specifications
| Antigen | Porimin |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | CL14381 |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | KCT-3, KCT3keratinocytes associated transmembrane protein 3, Keratinocytes-associated transmembrane protein 3, PORIMINporimin, pro oncosis receptor inducing membrane injury, Pro-oncosis receptor inducing membrane injury, serine/threonine-rich receptor, transmembrane protein 123PORMIN |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from Q8N131, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?