Learn More
Porimin Antibody (CL14381) [Alexa Fluor™ 700], Novus Biologicals™
Product Code. 30300619 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30300619 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30300619 Supplier Novus Biologicals Supplier No. NBP326852AF700
Mouse Monoclonal Antibody
Porimin Monoclonal antibody specifically detects Porimin in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Antigen Porimin
Applications Immunohistochemistry
Classification Monoclonal
Clone CL14381
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias KCT-3, KCT3keratinocytes associated transmembrane protein 3, Keratinocytes-associated transmembrane protein 3, PORIMINporimin, pro oncosis receptor inducing membrane injury, Pro-oncosis receptor inducing membrane injury, serine/threonine-rich receptor, transmembrane protein 123PORMIN
Host Species Mouse
Immunogen This antibody was developed using a synthetic peptide derived from Q8N131, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 114908
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG2b